CAS RN 1701-24-2
IUPAC Name:
4-chloro-2-(trifluoromethyl)quinoline
Synonyms:
4-chloro-2-(trifluoromethyl)quinoline
Molecular Weight (g/mol):
231.6
Molecular Formula:
C10H5ClF3N
InChi Key:
ONNDFDQMHCNEGF-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC(Cl)=C2C=CC=CC2=N1
1
–
1
of
1
results
1
4-Chloro-2-(trifluoromethyl)quinoline, 98%, Thermo Scientific™
CAS: 1701-24-2 Molecular Formula: C10H5ClF3N Molecular Weight (g/mol): 231.602 MDL Number: MFCD00153105 InChI Key: ONNDFDQMHCNEGF-UHFFFAOYSA-N Synonym: 4-chloro-2-trifluoromethyl quinoline,2-trifluoromethyl-4-chloroquinoline,4-chloro-2-trifluoromethyl-quinoline,quinoline, 4-chloro-2-trifluoromethyl,pubchem5854,buttpark 29\04-05,acmc-209e0j PubChem CID: 2736709 IUPAC Name: 4-chloro-2-(trifluoromethyl)quinoline SMILES: C1=CC=C2C(=C1)C(=CC(=N2)C(F)(F)F)Cl