accessibility menu, dialog, popup

UserName

CAS RN 16986-18-8

KKBrBrBrTeBrBrBr

CAS RN 16986-18-8

IUPAC Name: dipotassium tetrabromo-λ⁴-tellane dibromide
Synonyms: dipotassium tellurium tetrabromide dibromide
Molecular Weight (g/mol): 685.22
Molecular Formula: Br6K2Te
InChi Key: HMFRPYOIJYYQFO-UHFFFAOYSA-L
SMILES: [K+].[K+].[Br-].[Br-].Br[Te](Br)(Br)Br
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results