CAS RN 1698055-85-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
ARS-1620, MedChemExpress
MedChemExpress ARS-1620 is an atropisomeric selective KRASG12C inhibitor with desirable pharmacokinetics.