CAS RN 1698-60-8
CAS RN 1698-60-8
IUPAC Name:
5-amino-4-chloro-2-phenyl-2,3-dihydropyridazin-3-one
Synonyms:
chloridazon
Molecular Weight (g/mol):
221.64
Molecular Formula:
C10H8ClN3O
InChi Key:
WYKYKTKDBLFHCY-UHFFFAOYSA-N
SMILES:
NC1=C(Cl)C(=O)N(N=C1)C1=CC=CC=C1
1
–
3
of
3
results
2
Chloridazon, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.