Learn More
Chloridazon in Acetonitrile 100μg/mL, Fisher Chemical™
BIOPHARMA
Product Code. 12962414 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. 12962414
Brand: Fisher Chemical P810590
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Packaging Ampoule(s)
CAS 1698-60-8
Molecular Formula C10H8ClN3O
Concentration 100 μg/mL
MDL Number MFCD00055402
UN Number 1648
InChI Key WYKYKTKDBLFHCY-UHFFFAOYSA-N
SMILES NC1=C(Cl)C(=O)N(N=C1)C1=CC=CC=C1
IUPAC Name 5-amino-4-chloro-2-phenylpyridazin-3-one
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 221.64
PubChem CID 15546
Quantity 1 mL
ChEBI CHEBI:81838
Chemical Name or Material Chloridazon
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution Chloridazon,5-amino-4-chloro-2-phenylpyridazine-3-(2H)-one,pyrazon 100ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.