accessibility menu, dialog, popup

UserName

H3CSOHONHOOCH3CH3H3C

CAS RN 16947-82-3

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-(ethylsulfanyl)propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-(ethylsulfanyl)propanoic acid
Molecular Weight (g/mol): 249.33
Molecular Formula: C10H19NO4S
InChi Key: IBCCMMVPGKVLAX-UHFFFAOYNA-N
SMILES: CCSCC(NC(=O)OC(C)(C)C)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results