accessibility menu, dialog, popup

UserName

CAS RN 169255-50-9

IHOIFFF

CAS RN 169255-50-9

IUPAC Name: 2,6-diiodo-4-(trifluoromethyl)phenol
Synonyms: 2,6-diiodo-4-(trifluoromethyl)phenol
Molecular Weight (g/mol): 413.9
Molecular Formula: C7H3F3I2O
InChi Key: PFTYBCBURDKLTH-UHFFFAOYSA-N
SMILES: OC1=C(I)C=C(C=C1I)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results