accessibility menu, dialog, popup

UserName

FFFFFFNaSb

CAS RN 16925-25-0

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 258.74
Molecular Formula: F6NaSb
InChi Key: ZJLVIRKMUXZEDU-UHFFFAOYSA-N
SMILES: [F].[F].[F].[F].[F].[F].[Na+].[Sb]
Grade 
  • (7)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (5)
  • (2)
Physical Form 
  • (2)
  • (3)
  • (2)
Quantity 
  • (3)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (7)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)