CAS RN 16879-02-0
CAS RN 16879-02-0
IUPAC Name:
6-chloro-1,2-dihydropyridin-2-one
Synonyms:
6-chloro-1H-pyridin-2-one
Molecular Weight (g/mol):
129.54
Molecular Formula:
C5H4ClNO
InChi Key:
CLNNBQDAAGDAHI-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC(=O)N1
1
–
2
of
2
results
1
2-Chloro-6-hydroxypyridine, 98%
CAS: 16879-02-0 Molecular Formula: C5H4ClNO Molecular Weight (g/mol): 129.543 MDL Number: MFCD00006278 InChI Key: CLNNBQDAAGDAHI-UHFFFAOYSA-N Synonym: 6-chloropyridin-2-ol,2-chloro-6-hydroxypyridine,6-chloro-2-pyridinol,6-chloro-2-hydroxypyridine,6-chloropyridin-2 1h-one,2 1h-pyridinone, 6-chloro,6-chloro-2 1h-pyridinone,6-chloro-2-pyridone,6-chloropyridn-2-ol,2 1h-pyridone, 6-chloro PubChem CID: 85621 IUPAC Name: 6-chloro-1H-pyridin-2-one SMILES: C1=CC(=O)NC(=C1)Cl