accessibility menu, dialog, popup

UserName

Fluoroboric Acid

Fluoroboric acid is an inorganic, corrosive, strong acid with the chemical formula [H+][BF4-]. Available in various quantities, it is used as a precursor to fluoroborate salts and as a catalyst for alkylation and polymerization reactions.
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (7)
Percent Purity 
  • (2)
Showing products from the Fluoroboric Acid category. View All Search Results

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (2)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (3)
  • (7)
  • (2)