accessibility menu, dialog, popup

UserName

OOBrNOO

CAS RN 16869-24-2

IUPAC Name: (4-nitrophenyl)methyl 2-bromoacetate
Synonyms: (4-nitrophenyl)methyl 2-bromoacetate
Molecular Weight (g/mol): 274.07
Molecular Formula: C9H8BrNO4
InChi Key: ADHFTAKIDKDGBV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(COC(=O)CBr)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results