accessibility menu, dialog, popup

UserName

NNNOOH

CAS RN 167626-64-4

IUPAC Name: 3-(1H-1,2,4-triazol-1-yl)benzoic acid
Synonyms: 3-(1,2,4-triazol-1-yl)benzoic acid
Molecular Weight (g/mol): 189.17
Molecular Formula: C9H7N3O2
InChi Key: SZKWCOCFEIVCAB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC=C1)N1C=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results