accessibility menu, dialog, popup

UserName

CAS RN 16752-77-5

CH3HNOONH3CSCH3(E)

CAS RN 16752-77-5

IUPAC Name: (E)-[1-(methylsulfanyl)ethylidene]amino N-methylcarbamate
Synonyms: methomyl
Molecular Weight (g/mol): 162.21
Molecular Formula: C5H10N2O2S
InChi Key: UHXUZOCRWCRNSJ-QPJJXVBHSA-N
SMILES: CNC(=O)O\N=C(/C)SC
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)