accessibility menu, dialog, popup

UserName

CAS RN 16733-97-4

LiC

CAS RN 16733-97-4

IUPAC Name: lithium(1+) cyclopenta-1,3-dien-1-ide
Synonyms: lithium(1+) cyclopenta-1,3-dien-1-ide
Molecular Weight (g/mol): 72.04
Molecular Formula: C5H5Li
InChi Key: CSOHRTAOCDVTQU-UHFFFAOYSA-N
SMILES: [Li+].C1C=CC=[C-]1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results