accessibility menu, dialog, popup

UserName

CAS RN 166948-50-1

IUPAC Name: 2-fluoro-4-(trifluoromethoxy)benzoyl chloride
Synonyms: 2-fluoro-4-(trifluoromethoxy)benzoyl chloride
Molecular Weight (g/mol): 242.55
Molecular Formula: C8H3ClF4O2
InChi Key: VDZYQIMWGWFOEH-UHFFFAOYSA-N
SMILES: FC1=C(C=CC(OC(F)(F)F)=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results