accessibility menu, dialog, popup

UserName

CAS RN 166663-25-8

OHOOHOHOOHOHOOOHOHOOHOOHOONNHNHNHNHNHN(S)(S)(S)CH3(S)(S)(R)(R)(R)(S)(R)(S)(S)CH3(S)(R)(S)CH3CH3HHHHH

CAS RN 166663-25-8

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:
Molecular Weight (g/mol) 
  • (3)
  • (2)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results