Learn More
Anidulafungin, MedChemExpress
Product Code. 16478427
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478427 5 mg 5mg
16475748 10 mg 10mg
16471907 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16478427 Supplier Medchem Express Supplier No. HY13553
MedChemExpress Anidulafungin is a new semisynthetic echinocandin with antifungal potency.
Anidulafungin is a new semisynthetic echinocandin with antifungal potency.
Specifications
Chemical Name or Material Anidulafungin
Physical Form Powder
CAS 166663-25-8
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C58H73N7O17
For Use With (Application) COVID-19-immunoregulation
Synonym LY303366
Solubility Information DMSO : ≥ 30 mg/mL (26.31 mM)
SMILES O[C@@H]1C[C@H](N2C1)C(N[C@@H]([C@H](O)[C@@H](O)C3=CC=C(O)C=C3)C(N[C@@H]([C@@H](C)O)C(N4[C@@H]([C@@H](O)[C@@H](C)C4)C(N[C@H](O)[C@H](O)C[C@H](NC(C5=CC=C(C6=CC=C(C7=CC=C(OCCCCC)C=C7)C=C6)C=C5)=O)C(N[C@@H]([C@H](O)C)C2=O)=O)=O)=O)=O)=O
Molecular Weight (g/mol) 1140.24
Formula Weight 1140.24
Percent Purity 95.0%
Grade Research