accessibility menu, dialog, popup

UserName

CAS RN 16652-36-1

IUPAC Name: 2-bromo-3-fluoropropanoic acid
Synonyms: 2-bromo-3-fluoropropanoic acid
Molecular Weight (g/mol): 170.97
Molecular Formula: C3H4BrFO2
InChi Key: FBPUCVAAEPPEMG-UHFFFAOYNA-N
SMILES: OC(=O)C(Br)CF
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results