accessibility menu, dialog, popup

UserName

CAS RN 166328-14-9

BOFFFK

CAS RN 166328-14-9

IUPAC Name: potassium trifluoro(furan-2-yl)boranuide
Synonyms: potassium trifluoro(furan-2-yl)boranuide
Molecular Weight (g/mol): 173.97
Molecular Formula: C4H3BF3KO
InChi Key: IXQFYHBNPWHCCH-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results