accessibility menu, dialog, popup

UserName

CAS RN 1662-01-7

NN

CAS RN 1662-01-7

IUPAC Name: 4,7-diphenyl-1,10-phenanthroline
Synonyms: 4,7-diphenyl-1,10-phenanthroline
Molecular Weight (g/mol): 332.41
Molecular Formula: C24H16N2
InChi Key: DHDHJYNTEFLIHY-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=C2C=CC3=C(C=CN=C3C2=NC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)