CAS RN 166023-30-9
CAS RN 166023-30-9
IUPAC Name:
(3S)-1-(tert-butoxy)-5-methyl-1-oxohexan-3-aminium
Synonyms:
(3S)-1-(tert-butoxy)-5-methyl-1-oxohexan-3-aminium
Molecular Weight (g/mol):
202.32
Molecular Formula:
C11H24NO2
InChi Key:
WBOPVTYXLFEVGP-VIFPVBQESA-O
SMILES:
CC(C)C[C@H]([NH3+])CC(=O)OC(C)(C)C
1
–
1
of
1
results
1
tert-Butyl (3S)-3-amino-5-methylhexanoate, 95%
CAS: 166023-30-9 Molecular Formula: C11H23NO2 Molecular Weight (g/mol): 201.31 MDL Number: MFCD01076260 InChI Key: WBOPVTYXLFEVGP-VIFPVBQESA-N Synonym: tert-butyl 3s-3-amino-5-methylhexanoate,s-tert-butyl 3-amino-5-methylhexanoate,hexanoic acid,3-amino-5-methyl-, 1,1-dimethylethyl ester, 3s,tert-butyl 3s-3-amino-5-methyl-hexanoate,3s-3-amino-5-methylhexanoic acid tert-butyl ester,hexanoic acid,3-amino-5-methyl-,1,1-dimethylethyl ester, 3s PubChem CID: 7021556 IUPAC Name: tert-butyl (3S)-3-amino-5-methylhexanoate SMILES: CC(C)CC(CC(=O)OC(C)(C)C)N