accessibility menu, dialog, popup

UserName

CAS RN 16588-16-2

H3COOClNOO

CAS RN 16588-16-2

IUPAC Name: ethyl 4-chloro-3-nitrobenzoate
Synonyms: ethyl 4-chloro-3-nitrobenzoate
Molecular Weight (g/mol): 229.62
Molecular Formula: C9H8ClNO4
InChi Key: BLNLZRQIUGDTAO-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results