accessibility menu, dialog, popup

UserName

CAS RN 165806-95-1

CH3SOONCF

CAS RN 165806-95-1

IUPAC Name: 1-[(4-fluorophenyl)(isocyano)methanesulfonyl]-4-methylbenzene
Synonyms: 1-[(4-fluorophenyl)(isocyano)methanesulfonyl]-4-methylbenzene
Molecular Weight (g/mol): 289.32
Molecular Formula: C15H12FNO2S
InChi Key: UXCQPEDHCCJBNL-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)C([N+]#[C-])C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results