accessibility menu, dialog, popup

UserName

CAS RN 165736-03-8

NSNH2

CAS RN 165736-03-8

IUPAC Name: 1-(2-phenyl-1,3-thiazol-4-yl)methanamine
Synonyms: 1-(2-phenyl-1,3-thiazol-4-yl)methanamine
Molecular Weight (g/mol): 190.26
Molecular Formula: C10H10N2S
InChi Key: PMFCMLGAPHZRTE-UHFFFAOYSA-N
SMILES: NCC1=CSC(=N1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results