accessibility menu, dialog, popup

UserName

CAS RN 165669-32-9

NSOOSOOCl

CAS RN 165669-32-9

IUPAC Name: 4-(pyrrolidine-1-sulfonyl)benzene-1-sulfonyl chloride
Synonyms: 4-(pyrrolidine-1-sulfonyl)benzenesulfonyl chloride
Molecular Weight (g/mol): 309.78
Molecular Formula: C10H12ClNO4S2
InChi Key: ZXYZKIFFWKXROK-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(C=C1)S(=O)(=O)N1CCCC1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results