accessibility menu, dialog, popup

UserName

CAS RN 1653-30-1

H3CCH3OH

CAS RN 1653-30-1

IUPAC Name: undecan-2-ol
Synonyms: 2-undecanol
Molecular Weight (g/mol): 172.31
Molecular Formula: C11H24O
InChi Key: XMUJIPOFTAHSOK-UHFFFAOYNA-N
SMILES: CCCCCCCCCC(C)O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)