accessibility menu, dialog, popup

UserName

CAS RN 16502-01-5

HClNHNH

CAS RN 16502-01-5

IUPAC Name: hydrogen 1H,2H,3H,4H,9H-pyrido[3,4-b]indole chloride
Synonyms: tryptoline
Molecular Weight (g/mol): 208.69
Molecular Formula: C11H13ClN2
InChi Key: PHLJRXUBLWEPCM-UHFFFAOYSA-N
SMILES: [H+].[Cl-].C1CC2=C(CN1)NC1=CC=CC=C21
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results