accessibility menu, dialog, popup

UserName

CH3MgBr

CAS RN 16466-97-0

IUPAC Name: magnesium(2+) prop-1-yn-1-ide bromide
Synonyms: magnesium(2+) prop-1-yn-1-ide bromide
Molecular Weight (g/mol): 143.27
Molecular Formula: C3H3BrMg
InChi Key: QBNOPZJAURRQCE-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].CC#[C-]
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results