accessibility menu, dialog, popup

UserName

OOSN

CAS RN 16441-28-4

IUPAC Name: 2-(2-phenyl-1,3-thiazol-4-yl)acetate
Synonyms: 2-(2-phenyl-1,3-thiazol-4-yl)acetate
Molecular Weight (g/mol): 218.25
Molecular Formula: C11H8NO2S
InChi Key: LYHDWKGJPJRCTG-UHFFFAOYSA-M
SMILES: [O-]C(=O)CC1=CSC(=N1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results