accessibility menu, dialog, popup

UserName

SFFFBr

CAS RN 1644-72-0

IUPAC Name: 1-bromo-2-[(trifluoromethyl)sulfanyl]benzene
Synonyms: 1-bromo-2-[(trifluoromethyl)sulfanyl]benzene
Molecular Weight (g/mol): 257.07
Molecular Formula: C7H4BrF3S
InChi Key: IWRJVJJQWGZKMT-UHFFFAOYSA-N
SMILES: FC(F)(F)SC1=CC=CC=C1Br
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)