accessibility menu, dialog, popup

UserName

CAS RN 163931-61-1

H3CNCH3H3CH3CSiFF

CAS RN 163931-61-1

IUPAC Name: difluorotriphenylsilanuide; tetrabutylazanium
Synonyms: difluorotriphenylsilanuide; tetrabutylammonium ion
Molecular Weight (g/mol): 539.87
Molecular Formula: C34H51F2NSi
InChi Key: RQBKGJOQACIQDG-UHFFFAOYSA-N
SMILES: CCCC[N+](CCCC)(CCCC)CCCC.F[Si-](F)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results