accessibility menu, dialog, popup

UserName

CAS RN 16357-83-8

IUPAC Name: 4-aminopyrimidine-5-carbaldehyde
Synonyms: 4-aminopyrimidine-5-carbaldehyde
Molecular Weight (g/mol): 123.12
Molecular Formula: C5H5N3O
InChi Key: PDZKNPQAPGGAFJ-UHFFFAOYSA-N
SMILES: NC1=NC=NC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results