accessibility menu, dialog, popup

UserName

ClClSOOCl

CAS RN 163295-70-3

IUPAC Name: (3,5-dichlorophenyl)methanesulfonyl chloride
Synonyms: (3,5-dichlorophenyl)methanesulfonyl chloride
Molecular Weight (g/mol): 259.53
Molecular Formula: C7H5Cl3O2S
InChi Key: GZMJRPMBVICCFL-UHFFFAOYSA-N
SMILES: ClC1=CC(CS(Cl)(=O)=O)=CC(Cl)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results