accessibility menu, dialog, popup

UserName

CH3OOHNOOH3COHOH

CAS RN 1632-19-5

IUPAC Name: 2,5-dimethyl 3,4-dihydroxy-1H-pyrrole-2,5-dicarboxylate
Synonyms: 2,5-dimethyl 3,4-dihydroxy-1H-pyrrole-2,5-dicarboxylate
Molecular Weight (g/mol): 215.16
Molecular Formula: C8H9NO6
InChi Key: QCPBGHYMHYWRHG-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(O)C(O)=C(N1)C(=O)OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results