accessibility menu, dialog, popup

UserName

CAS RN 162607-18-3

BSClHOOH

CAS RN 162607-18-3

IUPAC Name: (5-chlorothiophen-2-yl)boronic acid
Synonyms: 5-chlorothiophen-2-ylboronic acid
Molecular Weight (g/mol): 162.39
Molecular Formula: C4H4BClO2S
InChi Key: JFCLNCVCDFUJPO-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(Cl)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results