accessibility menu, dialog, popup

UserName

CAS RN 1624261-27-3

ONHHCl

CAS RN 1624261-27-3

IUPAC Name: 3-phenylmorpholine hydrochloride
Synonyms: 3-phenylmorpholine hydrochloride
Molecular Weight (g/mol): 199.68
Molecular Formula: C10H14ClNO
InChi Key: BZPHPXWWVHPSOP-UHFFFAOYNA-N
SMILES: Cl.C1COCC(N1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results