accessibility menu, dialog, popup

UserName

CAS RN 162412-87-5

IUPAC Name: λ²-iron(2+) bis(1-(2,5-dimethylphospholan-1-yl)cyclopenta-2,4-dien-1-ide)
Synonyms: λ²-iron(2+) bis(1-(2,5-dimethylphospholan-1-yl)cyclopenta-2,4-dien-1-ide)
Molecular Weight (g/mol): 414.29
Molecular Formula: C22H32FeP2
InChi Key: QXXSYUGLJIMLFH-UHFFFAOYNA-N
SMILES: [Fe++].CC1CCC(C)P1[C-]1C=CC=C1.CC1CCC(C)P1[C-]1C=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results