accessibility menu, dialog, popup

UserName

CAS RN 1623-95-6

ClOO

CAS RN 1623-95-6

IUPAC Name: 4-phenoxybenzoyl chloride
Synonyms: 4-phenoxybenzoyl chloride
Molecular Weight (g/mol): 232.66
Molecular Formula: C13H9ClO2
InChi Key: AOOZVQGGMFGGEE-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results