accessibility menu, dialog, popup

UserName

CAS RN 16185-96-9

CH3OFFF

CAS RN 16185-96-9

IUPAC Name: 1-[2-(trifluoromethyl)phenyl]propan-1-one
Synonyms: 1-[2-(trifluoromethyl)phenyl]propan-1-one
Molecular Weight (g/mol): 202.18
Molecular Formula: C10H9F3O
InChi Key: PUSBIOFSWWHNDD-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results