CAS RN 161671-34-7
CAS RN 161671-34-7
IUPAC Name:
(1R)-3-(tert-butoxy)-3-oxo-1-phenylpropan-1-aminium
Synonyms:
(1R)-3-(tert-butoxy)-3-oxo-1-phenylpropan-1-aminium
Molecular Weight (g/mol):
222.31
Molecular Formula:
C13H20NO2
InChi Key:
TYYCBAISLMKLMT-LLVKDONJSA-O
SMILES:
CC(C)(C)OC(=O)C[C@@H]([NH3+])C1=CC=CC=C1
No Refinements
1
–
0
of
0
results
1
tert-Butyl (3R)-3-amino-3-phenylpropanoate, 97%
CAS: 161671-34-7 Molecular Weight (g/mol): 221.3 MDL Number: MFCD00798309 InChI Key: TYYCBAISLMKLMT-LLVKDONJSA-N Synonym: tert-butyl 3r-3-amino-3-phenylpropanoate,r-tert-butyl 3-amino-3-phenylpropanoate,3r-3-phenyl-beta-alanine tert-butyl ester,1,1-dimethylethyl 3r-3-amino-3-phenylpropanoate,tert-butyl-3r-3-amino-3-phenylpropanoate,benzenepropanoic acid,b-amino-,1,1-dimethylethyl ester, br,benzenepropanoic acid, b-amino-, 1,1-dimethylethyl ester, br PubChem CID: 2733824 IUPAC Name: tert-butyl (3R)-3-amino-3-phenylpropanoate SMILES: CC(C)(C)OC(=O)CC(C1=CC=CC=C1)N