CAS RN 16156-58-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Propargyl Methanesulfonate Ester, 95%, TRC
CAS: 16156-58-4 Molecular Formula: C4 H6 O3 S Molecular Weight (g/mol): 134.15 IUPAC Name: prop-2-ynyl methanesulfonate SMILES: CS(=O)(=O)OCC#C