CAS RN 16133-49-6
CAS RN 16133-49-6
IUPAC Name:
5-methoxy-2-nitroaniline
Synonyms:
3-amino-4-nitro-anisol
Molecular Weight (g/mol):
168.15
Molecular Formula:
C7H8N2O3
InChi Key:
QEHVRGACCVLLNN-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C(N)=C1)[N+]([O-])=O
1
–
1
of
1
results
1
5-Methoxy-2-nitroaniline, 98%
CAS: 16133-49-6 Molecular Formula: C7H8N2O3 Molecular Weight (g/mol): 168.152 MDL Number: MFCD00179573 InChI Key: QEHVRGACCVLLNN-UHFFFAOYSA-N Synonym: 5-methoxy-2-nitrophenylamine,3-amino-4-nitroanisole,5methoxy-2-nitroaniline,5-methoxy-2-nitrobenzenamine,3-amino-4-nitro-anisol,2-nitro-5-methoxyaniline,5-methoxy-2-nitro-aniline,benzenamine, 5-methoxy-2-nitro,benzenamine,5-methoxy-2-nitro,5-methoxy-2-nitro-phenylamine PubChem CID: 85300 IUPAC Name: 5-methoxy-2-nitroaniline SMILES: COC1=CC(=C(C=C1)[N+](=O)[O-])N