accessibility menu, dialog, popup

UserName

H3CCH3H2NNH2CH3CH3

CAS RN 16121-92-9

IUPAC Name: (2-methylpropyl)({6-[(2-methylpropyl)azaniumyl]hexyl})azanium
Synonyms: (2-methylpropyl)({6-[(2-methylpropyl)ammonio]hexyl})azanium
Molecular Weight (g/mol): 230.44
Molecular Formula: C14H34N2
InChi Key: LTRQCWUIJWVNNK-UHFFFAOYSA-P
SMILES: CC(C)C[NH2+]CCCCCC[NH2+]CC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results