accessibility menu, dialog, popup

UserName

CAS RN 16114-53-7

CH3NOI

CAS RN 16114-53-7

IUPAC Name: 4-iodo-3-methyl-5-phenyl-1,2-oxazole
Synonyms: 4-iodo-3-methyl-5-phenyl-1,2-oxazole
Molecular Weight (g/mol): 285.08
Molecular Formula: C10H8INO
InChi Key: RLZGZJOMCXKYIW-UHFFFAOYSA-N
SMILES: CC1=NOC(=C1I)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results