accessibility menu, dialog, popup

UserName

CAS RN 1611-03-6

CH3H3CCH3OOHOH3CH3CCH3

CAS RN 1611-03-6

IUPAC Name: 2-(3,5-di-tert-butyl-4-hydroxyphenyl)acetate
Synonyms: 2-(3,5-di-tert-butyl-4-hydroxyphenyl)acetate
Molecular Weight (g/mol): 263.36
Molecular Formula: C16H23O3
InChi Key: QLMGIWHWWWXXME-UHFFFAOYSA-M
SMILES: CC(C)(C)C1=CC(CC([O-])=O)=CC(=C1O)C(C)(C)C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)