CAS RN 16104-97-5
CAS RN 16104-97-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethylene Terephthalate Cyclic Pentamer, TRC
CAS: 16104-97-5 Molecular Formula: C50 H40 O20 Molecular Weight (g/mol): 960.841 Synonym: 3,6,13,16,23,26,33,36,43,46-Decaoxahexacyclo[46.2.2.28,11.218,21.228,31.238,41]hexaconta-8,10,18,20,28,30,38,40,48,50,51,53,55,57,59-pentadecaene-2,7,12,17,22,27,32,37,42,47-decone,Terephthalic Acid, Pentamol. Cyclic Ethylene Ester,Ethylene Glycol, Pentamol. Cyclic Terephthalate SMILES: O=C1OCCOC(=O)c2ccc(cc2)C(=O)OCCOC(=O)c3ccc(cc3)C(=O)OCCOC(=O)c4ccc(cc4)C(=O)OCCOC(=O)c5ccc(cc5)C(=O)OCCOC(=O)c6ccc1cc6