accessibility menu, dialog, popup

UserName

NNFFFH2N

CAS RN 16075-42-6

IUPAC Name: 4-(trifluoromethyl)pyrimidin-2-amine
Synonyms: 4-(trifluoromethyl)pyrimidin-2-amine
Molecular Weight (g/mol): 163.1
Molecular Formula: C5H4F3N3
InChi Key: NKOTXYPTXKUCDL-UHFFFAOYSA-N
SMILES: NC1=NC=CC(=N1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results