accessibility menu, dialog, popup

UserName

CAS RN 16066-91-4

IHN

CAS RN 16066-91-4

IUPAC Name: 5-iodo-1H-indole
Synonyms: 5-iodo-1H-indole
Molecular Weight (g/mol): 243.05
Molecular Formula: C8H6IN
InChi Key: TVQLYTUWUQMGMP-UHFFFAOYSA-N
SMILES: IC1=CC=C2NC=CC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)