accessibility menu, dialog, popup

UserName

CAS RN 16064-08-7

IONNH

CAS RN 16064-08-7

IUPAC Name: 6-iodo-1,4-dihydroquinazolin-4-one
Synonyms: 6-iodo-1H-quinazolin-4-one
Molecular Weight (g/mol): 272.05
Molecular Formula: C8H5IN2O
InChi Key: PUGXMZKDRVGIHC-UHFFFAOYSA-N
SMILES: IC1=CC=C2NC=NC(=O)C2=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results