accessibility menu, dialog, popup

UserName

CAS RN 1605-53-4

CH3PCH3

CAS RN 1605-53-4

IUPAC Name: diethyl(phenyl)phosphane
Synonyms: diethyl(phenyl)phosphane
Molecular Weight (g/mol): 166.2
Molecular Formula: C10H15P
InChi Key: LVTCZSBUROAWTE-UHFFFAOYSA-N
SMILES: CCP(CC)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results